Learn More
Mopec Ensemble Magnifier with LED Light
SDP
Catalog No. 23987039 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Magnifier with Light
Catalog No. For Use With (Equipment)
23-987-039 Magnifier with Light
1 options
Catalog No. 23-987-039 Supplier Mopec Supplier No. AX144
Add to Cart
Add to Cart
Designed to mount on the shelf to help better see specimens at a 1.75 x magnification.
  • Designed to mount on the shelf of a Maestro or Maestro Encore grossing station to help better see specimens at a 1.75 x magnification
  • Powder coated die cast aluminum
  • Magnifies 1.75 x
  • Light has color of 4000 K, 700 lumens, is dimmable, and has a 50,000-hour lamp life
  • Arm has 30 in.of extension and 180° of sweep
  • Fits all Mopec Maestro and Maestro Encore grossing stations
Specifications
Product Type Grossing Station
For Use With (Equipment) Magnifier with Light