Learn More
Mopec™ Ensemble Portable Vented Trash Can, Right
SDP
Catalog No. 22011114 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Portable Vented Trash Can
Catalog No. Product Type
22-011-114 Portable Vented Trash Can
1 options
Catalog No. 22-011-114 Supplier Mopec™ Supplier No. AX060R
Add to Cart
Add to Cart
Provides a vented trash receptacle solution next to your unit’s sink.
NOTE: This option must be ordered and at the time of purchase and installed during manufacturing of the grossing station. It cannot be installed in the field after delivery.
Specifications
Product Type Portable Vented Trash Can
Material 304 stainless steel construction with a polished No. 4 finish
Description Capacity; 60 lb.
Includes Trash can assembly on casters, flex duct, (2) clamps, and duct stub in side panel of grossing station unit
For Use With (Equipment) AX060-R is designed for right-handed Maestro and Maestro Encore grossing stations