Learn More
Medchemexpress LLC HY-143216 5mg , Ensitrelvir CAS:2647530-73-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1432165MG
Encompass_Preferred
HY-143216 5mg Ensitrelvir CAS: 2647530-73-0 Ensitrelvir (S-217622) is the first orally active non-covalent, non-peptidic, SARS-CoV-2 3CL protease inhibitor (IC50=13 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1849