Learn More
Selleck Chemical LLC Ensitrelvir fumarate-10MM/1ML
SDP
Catalog No. 503134652
Encompass_Preferred
S-217622 (Ensitrelvir fumarate) is the first orally active non-covalent non-peptidic SARS-CoV-2 3CL protease inhibitor with IC50 of 13 nM
Catalog No. 50-313-4652 Supplier Selleck Chemical LLC Supplier No. E113110MM/1ML
S-217622 (Ensitrelvir fumarate) is the first orally active non-covalent non-peptidic SARS-CoV-2 3CL protease inhibitor with IC50 of 13 nM