Learn More
Medchemexpress LLC Ensitrelvir fumarate | 2757470-18-9 | 100.0% | 647.95 g/mol | C26H21ClF3N9O6 | 5 MG
SDP
Catalog No. 5000221141
Encompass_Preferred
Ensitrelvir fumarate is the fumarate salt of an orally active, non-covalent, non-peptidic inhibitor of the SARS-CoV-2 3CL (main) protease. It is supplied as a high-purity solid for research use in antiviral, enzymatic, and mechanism-of-action studies.
  • Potent 3CL protease inhibition (IC50 13 nM).
  • Provided as the fumarate salt for improved handling and stability.
  • High purity appropriate for biochemical and cell-based assays.
  • Solid, white to off-white form for accurate weighing and formulation.
  • Small research pack sizes suitable for assay development and screening.
Catalog No. 50-002-21141 Supplier Medchemexpress LLC Supplier No. HY143216A5MG
Add to Cart
Add to Cart
Ensitrelvir fumarate is the fumarate salt of an orally active, non-covalent, non-peptidic inhibitor of the SARS-CoV-2 3CL (main) protease. It is supplied as a high-purity solid for research use in antiviral, enzymatic, and mechanism-of-action studies.
  • Potent 3CL protease inhibition (IC50 13 nM).
  • Provided as the fumarate salt for improved handling and stability.
  • High purity appropriate for biochemical and cell-based assays.
  • Solid, white to off-white form for accurate weighing and formulation.
  • Small research pack sizes suitable for assay development and screening.