Learn More
Medchemexpress LLC Ensitrelvir fumarate | 2757470-18-9 | 99.99% | C26H21ClF3N9O6 | 25 MG
SDP
Catalog No. 5000355904
Encompass_Preferred
Ensitrelvir fumarate is an orally active, non-covalent, and non-peptidic SARS-CoV-2 3CL protease inhibitor. It demonstrates significant inhibitory activity against various SARS-CoV-2 variants and has shown to inhibit intrapulmonary replication of SARS-CoV-2 in mice.
  • First orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor
  • Shows EC50 values of approximately 0.4 μM for wild-type and Alpha, Beta, Gamma, and Delta SARS-CoV-2 variants
  • Inhibits intrapulmonary replication of SARS-CoV-2 in mice
  • For research use only
Catalog No. 50-003-55904 Supplier Medchemexpress LLC Supplier No. 25MGHY143216A
Ensitrelvir fumarate is an orally active, non-covalent, and non-peptidic SARS-CoV-2 3CL protease inhibitor. It demonstrates significant inhibitory activity against various SARS-CoV-2 variants and has shown to inhibit intrapulmonary replication of SARS-CoV-2 in mice.
  • First orally active non-covalent, non-peptidic SARS-CoV-2 3CL protease inhibitor
  • Shows EC50 values of approximately 0.4 μM for wild-type and Alpha, Beta, Gamma, and Delta SARS-CoV-2 variants
  • Inhibits intrapulmonary replication of SARS-CoV-2 in mice
  • For research use only