Learn More
Enviro Tech International Ensolv Brand n-Propyl Bromide (nPB), 1 gal
SDP
Catalog No. NC1751230
Encompass
N-propyl bromide and therefore EnSolv offers a high efficacy of cleaning due to cyclical nature of vapor degreasing and also offers low global warming and ozone depletion potential of n propyl bromide.
Catalog No. NC1751230 Supplier Enviro Tech International Supplier No. ENO15408
N-propyl bromide and therefore EnSolv offers a high efficacy of cleaning due to cyclical nature of vapor degreasing and also offers low global warming and ozone depletion potential of n propyl bromide.