Learn More
Medchemexpress LLC Ensulizole | 27503-81-7 | 99.1% | 274.30 | 10 MG
SDP
Catalog No. 5000345048
Encompass_Preferred
Ensulizole is a sulfonated UV absorber that exhibits intense UVB and partial UVA absorption. It can damage DNA through the generation of reactive oxygen species (ROS) upon UV or sunlight irradiation. This product is for research use only.
  • Sulfonated UV absorber
  • Exhibits intense UVB and partial UVA absorption
  • Generates reactive oxygen species (ROS)
  • Solid appearance
  • White to yellow color
Catalog No. 50-003-45048 Supplier Medchemexpress LLC Supplier No. HY10965410G
Add to Cart
Add to Cart
Ensulizole is a sulfonated UV absorber that exhibits intense UVB and partial UVA absorption. It can damage DNA through the generation of reactive oxygen species (ROS) upon UV or sunlight irradiation. This product is for research use only.
  • Sulfonated UV absorber
  • Exhibits intense UVB and partial UVA absorption
  • Generates reactive oxygen species (ROS)
  • Solid appearance
  • White to yellow color