Learn More
Medchemexpress LLC HY-90009C 5mg Medchemexpress, ent-Tadalafil CAS:629652-72-8 Purity:>98%
SDP
Catalog No. 501970086
Encompass_Preferred
Medchemexpress, HY-90009C 5mg ent-Tadalafil CAS:629652-72-8 ent-Tadalafil (ent-IC-351), compound (6S,12aS), is a inactive cis-enantiomer of compound (6R,12aS). compound (6R,12aS) is a potent PDE5 inhibitor with an IC50 of 0.090 μM, while ent-Tadalafil is inactive at concentrations up to 10 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0086 Supplier Medchemexpress LLC Supplier No. HY90009C5MG
Medchemexpress, HY-90009C 5mg ent-Tadalafil CAS:629652-72-8 ent-Tadalafil (ent-IC-351), compound (6S,12aS), is a inactive cis-enantiomer of compound (6R,12aS). compound (6R,12aS) is a potent PDE5 inhibitor with an IC50 of 0.090 μM, while ent-Tadalafil is inactive at concentrations up to 10 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.