Learn More
TARGETMOL CHEMICALS INC ENTACAPONE 100MG
SDP
Catalog No. 502580973
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Entacapone (OR-611) is a selective and reversible inhibitor of catechol-O-methyltransferase (COMT). purity: 99%
Catalog No. 50-258-0973 Supplier TARGETMOL CHEMICALS INC Supplier No. T2216100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Entacapone (OR-611) is a selective and reversible inhibitor of catechol-O-methyltransferase (COMT). purity: 99%