Learn More
Apexbio Technology LLC Entacapone 130929-57-6 10mg
SDP
Catalog No. 5000096971
Encompass_Preferred
Entacapone (CAS 130929-57-6) is a selective second-generation inhibitor of catechol-O-methyltransferase (COMT) It demonstrates potent inhibition of total COMT in mouse liver with reported IC50 and Ki values of 20 1 nM and 10 7 nM respectively In rat models IC50 values for soluble COMT and membrane-bound COMT are 14 3 nM and 73 3 nM Entacapone is primarily utilized in research as an adjunct to levodopa and dopa decarboxylase inhibitor regimens in Parkinson s disease studies given its ability to suppress the formation of 3-O-methyldopa thereby enhancing levodopa bioavailability and prolonging its pharmacological effect
Catalog No. 50-000-96971 Supplier Apexbio Technology LLC Supplier No. A416810
Entacapone (CAS 130929-57-6) is a selective second-generation inhibitor of catechol-O-methyltransferase (COMT) It demonstrates potent inhibition of total COMT in mouse liver with reported IC50 and Ki values of 20 1 nM and 10 7 nM respectively In rat models IC50 values for soluble COMT and membrane-bound COMT are 14 3 nM and 73 3 nM Entacapone is primarily utilized in research as an adjunct to levodopa and dopa decarboxylase inhibitor regimens in Parkinson s disease studies given its ability to suppress the formation of 3-O-methyldopa thereby enhancing levodopa bioavailability and prolonging its pharmacological effect