Learn More
Medchemexpress LLC Entacapone | 130929-57-6 | MFCD00866580 | 100.0% | C14H15N3O5 | 10MG
SDP
Catalog No. 5000260311
Encompass_Preferred
Entacapone is a potent, reversible, peripherally acting catechol-O-methyltransferase (COMT) inhibitor provided as an analytical research standard for in vitro and biochemical studies, including Parkinson's disease and metabolic enzyme research. The material is supplied as a solid or solution and is intended for research use only.
  • High purity for analytical and assay applications.
  • Validated for COMT inhibition studies.
  • Available in multiple small-scale pack sizes for laboratory use.
  • Offered as both solid and premade solution formats.
  • Useful as a reference standard in biochemical and enzymatic assays.
Catalog No. 50-002-60311 Supplier Medchemexpress LLC Supplier No. HY1428010MG
Entacapone is a potent, reversible, peripherally acting catechol-O-methyltransferase (COMT) inhibitor provided as an analytical research standard for in vitro and biochemical studies, including Parkinson's disease and metabolic enzyme research. The material is supplied as a solid or solution and is intended for research use only.
  • High purity for analytical and assay applications.
  • Validated for COMT inhibition studies.
  • Available in multiple small-scale pack sizes for laboratory use.
  • Offered as both solid and premade solution formats.
  • Useful as a reference standard in biochemical and enzymatic assays.