Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Entacapone | 130929-57-6 | MFCD00866580 | >98.0% | 305.29 g/mol | C14H15N3O5 | 5 MG
SDP

Catalog No. 5000220996
Encompass_Preferred

Entacapone is a research-grade catechol-O-methyltransferase (COMT) inhibitor supplied as a small-scale reagent. It is potent, reversible, peripherally acting, and orally active, and is used in Parkinson's disease research and studies of FTO demethylation.

  • Potent, reversible COMT inhibitor.
  • Peripherally acting and orally active.
  • Suitable for Parkinson's disease research.
  • Inhibits FTO demethylation for metabolic research.
  • Purity >98.0% (HPLC) reported by suppliers.
  • Molecular formula C14H15N3O5; molecular weight 305.29 g/mol.
  • Supplied as a small-scale research pack (5 mg).

Catalog No. 50-002-20996 Supplier Medchemexpress LLC Supplier No. HY142805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Entacapone is a research-grade catechol-O-methyltransferase (COMT) inhibitor supplied as a small-scale reagent. It is potent, reversible, peripherally acting, and orally active, and is used in Parkinson's disease research and studies of FTO demethylation.

  • Potent, reversible COMT inhibitor.
  • Peripherally acting and orally active.
  • Suitable for Parkinson's disease research.
  • Inhibits FTO demethylation for metabolic research.
  • Purity >98.0% (HPLC) reported by suppliers.
  • Molecular formula C14H15N3O5; molecular weight 305.29 g/mol.
  • Supplied as a small-scale research pack (5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.