Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Entacapone | 130929-57-6 | MFCD00866580 | 99.2% | 305.29 g/mol | C14H15N3O5 | 5 MG
SDP

Catalog No. 5000221367
Encompass_Preferred

Entacapone (Standard) is an analytical reference standard of entacapone provided for research and analytical use. Supplied in small quantitative sizes suitable for assay development, method validation, and quality control, the product includes batch documentation and a Certificate of Analysis reporting high purity. It is used as a reference material in studies involving COMT inhibition and related biochemical assays.

  • Analytical standard for assay development and method validation.
  • Available in small sizes, including a 5 mg vial.
  • Certified purity approximately 99.2% (COA available).
  • Useful for quality control and reference material applications.
  • Molecular formula C14H15N3O5; molecular weight 305.29 g/mol.

Catalog No. 50-002-21367 Supplier Medchemexpress LLC Supplier No. HY14280R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Entacapone (Standard) is an analytical reference standard of entacapone provided for research and analytical use. Supplied in small quantitative sizes suitable for assay development, method validation, and quality control, the product includes batch documentation and a Certificate of Analysis reporting high purity. It is used as a reference material in studies involving COMT inhibition and related biochemical assays.

  • Analytical standard for assay development and method validation.
  • Available in small sizes, including a 5 mg vial.
  • Certified purity approximately 99.2% (COA available).
  • Useful for quality control and reference material applications.
  • Molecular formula C14H15N3O5; molecular weight 305.29 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.