Learn More
Medchemexpress LLC Entacapone | 130929-57-6 | MFCD00866580 | 99.2% | C14H15N3O5 | 10MG
SDP
Catalog No. 5000261874
Encompass_Preferred
Entacapone (Standard) is an analytical reference standard of entacapone (CAS 130929-57-6) supplied for research and analytical applications. It is intended for use in qualitative and quantitative assays, method development, and validation for chromatographic and mass spectrometric techniques.
  • Provides a certified analytical standard for assay development and method validation.
  • High purity reference material (≈99.2%).
  • Suitable for HPLC, GC, and MS applications.
  • Light yellow solid, stable under recommended storage conditions.
  • Supports studies of catechol-O-methyltransferase activity and Parkinson's disease research.
Catalog No. 50-002-61874 Supplier Medchemexpress LLC Supplier No. HY14280R10MG
Entacapone (Standard) is an analytical reference standard of entacapone (CAS 130929-57-6) supplied for research and analytical applications. It is intended for use in qualitative and quantitative assays, method development, and validation for chromatographic and mass spectrometric techniques.
  • Provides a certified analytical standard for assay development and method validation.
  • High purity reference material (≈99.2%).
  • Suitable for HPLC, GC, and MS applications.
  • Light yellow solid, stable under recommended storage conditions.
  • Supports studies of catechol-O-methyltransferase activity and Parkinson's disease research.