Learn More
Medchemexpress LLC Entacapone (Standard) | 130929-57-6 | 99.2% | C14H15N3O5 | 50 MG
SDP
Catalog No. 5000354117
Encompass_Preferred
Entacapone (Standard) is the analytical standard of Entacapone. It is a potent, reversible, peripherally acting, and orally active catechol-O-methyltransferase (COMT) inhibitor. It inhibits COMT from rat brain, erythrocytes, and liver with IC50 values of 10 nM, 20 nM, and 160 nM, respectively. It is selective for COMT over other catecholamine metabolizing enzymes, including MAO-A, MAO-B, phenolsulphotransferase M (PST-M), and PST-P (IC50s>50 μM).
  • Used for research of Parkinson's disease
  • Serves as an inhibitor of FTO demethylation with an IC50 of 3.5 μM, for research of metabolic disorders
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Intended for research and analytical applications
Catalog No. 50-003-54117 Supplier Medchemexpress LLC Supplier No. HY14280R50MG
Entacapone (Standard) is the analytical standard of Entacapone. It is a potent, reversible, peripherally acting, and orally active catechol-O-methyltransferase (COMT) inhibitor. It inhibits COMT from rat brain, erythrocytes, and liver with IC50 values of 10 nM, 20 nM, and 160 nM, respectively. It is selective for COMT over other catecholamine metabolizing enzymes, including MAO-A, MAO-B, phenolsulphotransferase M (PST-M), and PST-P (IC50s>50 μM).
  • Used for research of Parkinson's disease
  • Serves as an inhibitor of FTO demethylation with an IC50 of 3.5 μM, for research of metabolic disorders
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Intended for research and analytical applications