Learn More
Cayman Chemical Entacaponed10 GC MS Standards
SDP
Catalog No. 502891386
Encompass_Preferred
An internal standard for the quantification of entacapone by GC- or LC-MS
Catalog No. 50-289-1386 Supplier Cayman Chemical Supplier No. 100105711 MG
Add to Cart
Add to Cart
An internal standard for the quantification of entacapone by GC- or LC-MS