Learn More
Cayman Chemical Entacaponed10 GC MS Standards
SDP
Catalog No. 502890665
Encompass_Preferred
An internal standard for the quantification of entacapone by GC- or LC-MS
Catalog No. 50-289-0665 Supplier Cayman Chemical Supplier No. 10010571500 UG
An internal standard for the quantification of entacapone by GC- or LC-MS