Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Entadamide-A-β-D-glucopyranoside | 138916-58-2 | 25 MG
SDP

Catalog No. 5000420430
Encompass_Preferred

Entadamide-A-β-D-glucopyranoside is a chemical compound and a major component found in the seeds of *Entada phaseoloides*. It is known for its anti-complement activities.

  • High purity of 98.15%
  • Molecular formula C12H21NO7S
  • Molecular weight 323.36
  • Appears as a white to off-white solid
  • Classified as an alkaloid
  • Derived from plants
  • Exhibits anti-complement activities

Catalog No. 50-004-20430 Supplier Medchemexpress LLC Supplier No. 25MGHYN7401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Entadamide-A-β-D-glucopyranoside is a chemical compound and a major component found in the seeds of *Entada phaseoloides*. It is known for its anti-complement activities.

  • High purity of 98.15%
  • Molecular formula C12H21NO7S
  • Molecular weight 323.36
  • Appears as a white to off-white solid
  • Classified as an alkaloid
  • Derived from plants
  • Exhibits anti-complement activities

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.