Learn More
ATCC Entamoeba histolytica; HM-1:IMSS [ABRM]; Quantitative DNA
Supplier:  ATCC 30459DQ
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load and for other molecular biology applications.
Catalog No. 50-238-5003