Learn More
Medchemexpress LLC Entasobulin | 501921-61-5 | 98.5% | 439.89 | 50 MG
SDP
Catalog No. 5000393864
Encompass_Preferred
Entasobulin is a β-tubulin polymerization inhibitor with potential anticancer activity. It is an indolizine-glyoxylamide based small molecule that demonstrates substantial in vitro anti-proliferative activities against cancer cell lines, including multidrug resistance (MDR) phenotypes.
  • Inhibits β-tubulin polymerization
  • Shows potential anticancer activity
  • Demonstrates substantial in vitro anti-proliferative activities against cancer cell lines
  • Effective against multidrug resistance (MDR) phenotypes
  • For research use only
Catalog No. 50-003-93864 Supplier Medchemexpress LLC Supplier No. HY1677750MG
Entasobulin is a β-tubulin polymerization inhibitor with potential anticancer activity. It is an indolizine-glyoxylamide based small molecule that demonstrates substantial in vitro anti-proliferative activities against cancer cell lines, including multidrug resistance (MDR) phenotypes.
  • Inhibits β-tubulin polymerization
  • Shows potential anticancer activity
  • Demonstrates substantial in vitro anti-proliferative activities against cancer cell lines
  • Effective against multidrug resistance (MDR) phenotypes
  • For research use only