Learn More
Cayman Chemical EntecavIr-d2 1mg
SDP
Catalog No. 502915425
Encompass_Preferred
An internal standard for the quantification of entecavir by GC- or LC-MS
Catalog No. 50-291-5425 Supplier Cayman Chemical Supplier No. 372121 MG
Add to Cart
Add to Cart
An internal standard for the quantification of entecavir by GC- or LC-MS