Learn More
Apexbio Technology LLC Entecavir Hydrate 209216-23-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063697
Encompass_Preferred
Entecavir hydrate is an antiviral compound that acts as a reverse transcriptase (RT) inhibitor specifically targeting hepatitis B virus (HBV) infection It inhibits HBV DNA replication by interacting with the viral reverse transcriptase enzyme thus preventing the synthesis of complementary DNA from the viral RNA template Entecavir hydrate is frequently employed in antiviral studies and clinical research related to hepatitis B treatment regimens In studies involving chronic HBV-infected patients entecavir hydrate combined with peginterferon alpha-2b therapy has shown significant suppression of viral DNA levels and reduced covalently closed circular DNA (cccDNA) within hepatocytes The reported IC50 value of entecavir hydrate against HBV DNA polymerase activity typically ranges below 10 nM in cellular assay systems enabling its utility in both preclinical and clinical antiviral investigations
Catalog No. 50-000-63697 Supplier Apexbio Technology LLC Supplier No. A17675.1
Entecavir hydrate is an antiviral compound that acts as a reverse transcriptase (RT) inhibitor specifically targeting hepatitis B virus (HBV) infection It inhibits HBV DNA replication by interacting with the viral reverse transcriptase enzyme thus preventing the synthesis of complementary DNA from the viral RNA template Entecavir hydrate is frequently employed in antiviral studies and clinical research related to hepatitis B treatment regimens In studies involving chronic HBV-infected patients entecavir hydrate combined with peginterferon alpha-2b therapy has shown significant suppression of viral DNA levels and reduced covalently closed circular DNA (cccDNA) within hepatocytes The reported IC50 value of entecavir hydrate against HBV DNA polymerase activity typically ranges below 10 nM in cellular assay systems enabling its utility in both preclinical and clinical antiviral investigations