Learn More
Apexbio Technology LLC Entecavir Hydrate, 25mg. Cas: 209216-23-9 MFCD: MFCD09754448. Antiviral drug used in hepatitis B infection
SDP
Catalog No. 501904707
Encompass_Preferred
Entecavir hydrate is a reverse transcriptase inhibitor.Reverse transcriptase (RT) is used to generate complementary DNA (cDNA) from an RNA template, which is mainly associated with retroviruses, such as HBV. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4707 Supplier Apexbio Technology LLC Supplier No. A176725
Entecavir hydrate is a reverse transcriptase inhibitor.Reverse transcriptase (RT) is used to generate complementary DNA (cDNA) from an RNA template, which is mainly associated with retroviruses, such as HBV. Other sizes are available. Please inqury us for quote.