Learn More
AdipoGen Entecavir monohydrate (200 mg)
SDP
Catalog No. 502054419
Encompass_Preferred
Chemical. CAS: 209216-23-9. Formula: C12H15N5O3 . H2O. MW: 295.29. Synthetic. Entecavir is an oral antiviral drug used in the treatment of hepatitis B (HBV) infection. It is marketed under the trade name Baraclude (BMS). Entecavir is a potent deoxyguanosine nucleoside analog with antiviral activity selective for hepadnaviruses. In vitro, the active intracellular form of entecavir, entecavir triphosphate, demonstrates a higher binding affinity for HBV DNA polymerase than the natural guanosine triphosphate substrate and effectively inhibits HBV DNA replication at 3 stages in the replication pathway: priming, reverse transcription and DNA-dependent DNA synthesis. Can be also used as a reference compound.
Catalog No. 50-205-4419 Supplier AdipoGen Supplier No. CDXE0229M200
Chemical. CAS: 209216-23-9. Formula: C12H15N5O3 . H2O. MW: 295.29. Synthetic. Entecavir is an oral antiviral drug used in the treatment of hepatitis B (HBV) infection. It is marketed under the trade name Baraclude (BMS). Entecavir is a potent deoxyguanosine nucleoside analog with antiviral activity selective for hepadnaviruses. In vitro, the active intracellular form of entecavir, entecavir triphosphate, demonstrates a higher binding affinity for HBV DNA polymerase than the natural guanosine triphosphate substrate and effectively inhibits HBV DNA replication at 3 stages in the replication pathway: priming, reverse transcription and DNA-dependent DNA synthesis. Can be also used as a reference compound.