Learn More
Medchemexpress LLC HY-13623A 200mg , Entecavir (monohydrate) SQ 34676;BMS-200475 CAS:209216-23-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13623A200MG
Encompass_Preferred
Medchemexpress, HY-13623A 200mg Entecavir (monohydrate) SQ 34676;BMS-200475 CAS:209216-23-9 Formula:C12H17N5O4 EC50:3.75 nM (anti-HBV, HepG2 cell) Purity:98% Entecavir monohydrate (SQ 34676; BMS 200475) is a potent and selective inhibitor of HBV , with an EC 50 of 3.75 nM in HepG2 cell. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2658