Learn More
Cayman Chemical Entecavir (hydrate), Purity: ≥95%, CAS: 209216-23-9, 100 mg
SDP
Catalog No. 502955280
Encompass_Preferred
An antiviral nucleoside analog and inhibitor of HBV reverse transcriptase (IC50 = 0.5 nM); reduces virion DNA in the culture supernatant of HepG2 2.2.15 cells infected with HBV (EC50 = 3.75 nM); reduces serum and hepatic levels of viral DNA in a duckling model of HBV infection at 1 mg/kg
Catalog No. 50-295-5280 Supplier Cayman Chemical Supplier No. 13831100 MG
An antiviral nucleoside analog and inhibitor of HBV reverse transcriptase (IC50 = 0.5 nM); reduces virion DNA in the culture supernatant of HepG2 2.2.15 cells infected with HBV (EC50 = 3.75 nM); reduces serum and hepatic levels of viral DNA in a duckling model of HBV infection at 1 mg/kg