Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Entegris 200mm Ultrapak Assemblies For Wafer Shipping Boxes

Catalog No. 19165267
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
200 mm Wafer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-165-267 200 mm Wafer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-165-267 Supplier Entegris Supplier No. H92000022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

For Use With (Equipment) 200 mm Wafer
Applications Clean and secure transport of 200 mm wafers
Material Wafershield™ high-purity polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.