Learn More
Electron Microscopy Sciences Entellan Rapid Mounting Media 100 ML
SDP
Catalog No. 50980442
Encompass_Preferred
Entellan is a rapid mounting media for microscopy. It can be used for long life preparations, without bubble formation at high ambient temperatures. Also for all dehydrated microscopic preparations. Cure time is 20 minutes at room temperature. It is colorless with an acid number less than 2.50 and a R.I. ND20 approx 1.49-1.5. (Contains xylene).
Catalog No. 50-980-442 Supplier Electron Microscopy Sciences Supplier No. 14800
Entellan is a rapid mounting media for microscopy. It can be used for long life preparations, without bubble formation at high ambient temperatures. Also for all dehydrated microscopic preparations. Cure time is 20 minutes at room temperature. It is colorless with an acid number less than 2.50 and a R.I. ND20 approx 1.49-1.5. (Contains xylene).