Learn More
MSC Re-enterable Encapsulant, 3M, 2-Part, Non-Urethane, 90 mL, 1 Each
Catalog No. 502870163
Encompass
The 8882 high gel re-enterable encapsulant is a 2-part, non-urethane compound that allows for easy cable splice re-entry for future changes and maintenance. It adheres to filled cables and is compatible with common plastics in telephone splice connectors, forming a reliable seal for outdoor and buried applications.
  • Enables easy cable splice re-entry for future changes and maintenance
  • Has 100 to 110 minute gel time at 40 degrees F (4 degrees C)
  • Adheres to filled cable and is compatible with common plastics used in telephone splice connectors
  • Two-part, non-urethane encapsulating compound
  • Ideal for outdoor and buried applications
  • Used to seal and protect copper connections inside an enclosure
  • Provides excellent thermal protection and can withstand elevated temperatures
Catalog No. 50-287-0163 Supplier MSC Supplier No. 89501167
The 8882 high gel re-enterable encapsulant is a 2-part, non-urethane compound that allows for easy cable splice re-entry for future changes and maintenance. It adheres to filled cables and is compatible with common plastics in telephone splice connectors, forming a reliable seal for outdoor and buried applications.
  • Enables easy cable splice re-entry for future changes and maintenance
  • Has 100 to 110 minute gel time at 40 degrees F (4 degrees C)
  • Adheres to filled cable and is compatible with common plastics used in telephone splice connectors
  • Two-part, non-urethane encapsulating compound
  • Ideal for outdoor and buried applications
  • Used to seal and protect copper connections inside an enclosure
  • Provides excellent thermal protection and can withstand elevated temperatures