Learn More
Med Vet International Enteral Feeding Products, J0350J, 10Fr x 55in, Each
Catalog No. 5000582944
Encompass_Preferred
This line of products is especially designed for mid-long term enteral feeding. Features of polyurethane feeding tubes: More durable and longer lasting than PVC tubes; Weighted tip with metal tungsten guide end for ease of placement and retention; Stylets included on all sizes; Radiopaque and non-sterile; Many of these tubes will move by peristaltic action to the jejunum after placement in the stomach, which requires longer lengths than standard feeding tubes; Numerical graduations on side of the tube; and Complete directions come with the product.
Catalog No. 50-005-82944 Supplier Med Vet International Supplier No. JOR350J
This line of products is especially designed for mid-long term enteral feeding. Features of polyurethane feeding tubes: More durable and longer lasting than PVC tubes; Weighted tip with metal tungsten guide end for ease of placement and retention; Stylets included on all sizes; Radiopaque and non-sterile; Many of these tubes will move by peristaltic action to the jejunum after placement in the stomach, which requires longer lengths than standard feeding tubes; Numerical graduations on side of the tube; and Complete directions come with the product.