Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Enteric Bacterial Verification Panel (Live Culture)
SDP

Catalog No. 23017294 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
16 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-017-294 16 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-294 Supplier Microbiologics™ Supplier No. 8179
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics offers a wide range of bundled QC Sets and Panels for the most commonly used technologies and standards. Each set or panel contains the manufacturer-recommended quality control strains for a particular instrument or test method in a simple, ready-to-use format.

Enteric Bacterial Verification Panel (Live Culture) includes two each of Campylobacter jejuni subsp. jejuni derived from ATCC™ 33291™, Campylobacter coli derived from ATCC™ 33559™, Salmonella enterica subsp. enterica serovar Typhimurium derived from ATCC™ 14028™, Salmonella bongori derived from ATCC™ 43975™, Shigella sonnei derived from ATCC™ 9290™,Shigella flexneri derived from ATCC™ 12022™,Escherichia coli (O157:H7) derived from ATCC™ 43890™ (STEC), and Escherichia coli serotype O111:H8 derived from CDC 2010C-3114 (STEC)

Specifications

Content And Storage 2°C to 8°C
Format QC Sets and Panels: KWIK-STIK™
Manufacturing Origin United States
Product Type Quality Control
No. per Pack 2
Packaging Type Plastic Canister
Quantity 16 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.