Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Enteric Parasite Control Panel (Inactivated Pellet)
SDP

Catalog No. 23004145 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-145 6 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-004-145 Supplier Microbiologics™ Supplier No. 8230
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enteric Parasite Control Panel (Inactivated Pellet) contains six inactivated, pre-pooled pellets comprised of the microorganisms.

Each pellet contains the microorganisms: Cryptosporidium parvum, Entamoeba histolytica surrogate, Giardia lamblia 

Specifications

Biosafety Level 1
Content And Storage 2 to 25°C
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) Microbial Identification, Molecular Testing
Packaging Type Plastic Box
Quantity 6 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.