Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Enteric Viral Control Panel
SDP

Catalog No. 23046868 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-868 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-046-868 Supplier Microbiologics™ Supplier No. 8211
Add to Cart
Add to Cart

Microbiologics™ Enteric Viral Control Panel includes six inactivated microorganism pellets each comprised of five organisms

  • Pellets are stable at room-temperature
  • For in vitro diagnostic use
  • Currently not available for shipment to Canada
  • Organisms:
    • Adenovirus 40
    • Recombinant Astrovirus
    • Recombinant Norovirus GII
    • Rotavirus
    • Recombinant Sapovirus

Specifications

Certifications/Compliance IVD, CE Marked
Content And Storage Room Temperature
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) In vitro diagnostics
Packaging Type Plastic Box
Quantity 6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.