Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Enteric Viral Verification Panel
SDP

Catalog No. 23046867 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-867 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-867 Supplier Microbiologics™ Supplier No. 8210
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics™ Enteric Viral Verification Panel includes 20 inactivated microorganism pellets each comprised of five organisms

  • Pellets are stable at room-temperature
  • For in vitro diagnostic use
  • Currently not available for shipment to Canada
  • Organisms:
    • Adenovirus 40
    • Recombinant Astrovirus
    • Recombinant Norovirus GII
    • Rotavirus
    • Recombinant Sapovirus

Specifications

Certifications/Compliance IVD, CE Marked
Content And Storage Room Temperature
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) In vitro diagnostics
Packaging Type Plastic Box
Quantity 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.