Learn More
Micro Biologics Inc 3P ENTEROBACTER AEROGENES 6PK
SDP
Catalog No. NC0365682
Encompass_Preferred
LYFO DISK® is a lyophilized pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option. LYFO DISK® is known for its ease-of-use; simply re-hydrate the pellet and inoculate. LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain. Enterobacter aerogenes derived from ATCC® 13048™*
Catalog No. NC0365682 Supplier Micro Biologics Inc 3P Supplier No. 0306L
LYFO DISK® is a lyophilized pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option. LYFO DISK® is known for its ease-of-use; simply re-hydrate the pellet and inoculate. LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain. Enterobacter aerogenes derived from ATCC® 13048™*