Learn More
Biotium Enterobacter aerogenes(57/82), CF594 conjugate, 0.1mg/mL
Supplier:  Biotium BNC940208500
Encompass_Preferred
Enterobacter aerogenesis a Gram-negative rod-shaped microorganism from the Enterobacteriaceae family. E. aerogenes forms part of the endogenous human gastrointestinal (GI) microflora. It also resides in soil, water and in dairy products. Generally infections arise from the patients -lactam antibiotics, aminoglycosides and quinolones. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-252-3345