Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Enterobacter cloacae derived from ATCC™ BAA-2341
SDP

Catalog No. 23045904 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-904 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-045-904 Supplier Microbiologics™ Supplier No. 01202P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics Enterobacter cloacae derived from ATCC™ BAA-2341 is a simple, all-inclusive design that makes life easier for lab techs while reducing chances for errors

  • Biosafety Level: 1
  • 2 self-contained units of a single organism; blaKPC positive, produces carbapenemase KPC, carbapenem-resistant (Imipenem and Ertapenem)
  • Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab

Specifications

ATCC® BAA-2341
Biosafety Level 2
Content And Storage 2°C to 8°C
Format KWIK-STIK
Manufacturing Origin United States
For Use With (Application) Microbial Identification
Packaging Type Plastic Canister
Quantity 2
Species Enterobacter cloacae
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.