Learn More
ATCC Enterobacter cloacae; Strain 1000654
Supplier:  ATCC BAA2468
Encompass
Enterobacter cloacae strain 1000654 is a whole-genome sequenced bacterium that was isolated from human urine. This strain is resistant to ertapenem and imipenem and was confirmed by PCR to contain the blaNDM gene. The strain can be used for antimicrobial resistance research, drug development, and infectious disease research.
Catalog No. 50-238-3151