Learn More
ATCC Enterobacter cloacae; Strain 1101152
Catalog No. 502385011
Encompass
Enterobacter cloacae strain 1101152 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This strain demonstrates resistance to ertapenem and imipenem and was confirmed by PCR to contain the blaKPC gene. It can be used in antimicrobial resistance research and multidrug testing.
Catalog No. 50-238-5011 Supplier ATCC Supplier No. BAA2341
Enterobacter cloacae strain 1101152 is a whole-genome sequenced bacterium that was isolated from a clinical specimen. This strain demonstrates resistance to ertapenem and imipenem and was confirmed by PCR to contain the blaKPC gene. It can be used in antimicrobial resistance research and multidrug testing.