Learn More
ATCC Enterobacter hormaechei subsp. steigerwaltii; Strain: SC10435
Catalog No. 502379888
Encompass
Enterobacter hormaechei subsp. steigerwaltii strain SC10435 was isolated in 1963 from human blood culture in Toronto, Canada. This strain is resistant to penicillins, cephalosporins, aztreonam, and pipercillin-tazobactam and it is sensitive to carbapenems. The depositor states that this strain produces a Class C, AmpC-type beta-lactamase (P99 cephalosporinase).
Catalog No. 50-237-9888 Supplier ATCC Supplier No. BAA2940
Enterobacter hormaechei subsp. steigerwaltii strain SC10435 was isolated in 1963 from human blood culture in Toronto, Canada. This strain is resistant to penicillins, cephalosporins, aztreonam, and pipercillin-tazobactam and it is sensitive to carbapenems. The depositor states that this strain produces a Class C, AmpC-type beta-lactamase (P99 cephalosporinase).