Learn More
ATCC Enterobacter hormaechei
Supplier:  ATCC 700323
Encompass
Enterobacter hormaechei strain LBM 93.03.067 is a whole-genome sequenced bacterium that can be used as a quality control for the Vitek 2 ID-GNB card.
Catalog No. 50-238-3701