Learn More
AdipoGen Enterocin (1 mg)
SDP
Supplier:  AdipoGen AGCN20116M001
Encompass_Preferred
Chemical. CAS: 59678-46-5. Formula: C22H20O10. MW: 444.4. Isolated from Streptomyces sp. Potent antibiotic. Bacteriocin. Antibacterial. Broad spectrum activity against Gram-positive and Gram-negative bacteria. Herbicidal. Enterocin is a small molecular weight secondary metabolite and should not be confused with bacterial proteins isolated from Enterococci generically referred to as "enterocins".
Catalog No. 50-205-3481