Learn More
MOLTOX Molecular Toxicology Inc Enterococcosel Agar Plate, 20 mL/plate, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948692
Encompass_Preferred
Also known as Bile Esculin Azide Agar, Enterococcosel Agar is used as a rapid selective medium for the detection and enumeration of enterococci. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-692 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S83
Also known as Bile Esculin Azide Agar, Enterococcosel Agar is used as a rapid selective medium for the detection and enumeration of enterococci. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.