Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC BAA2365
Encompass
Enterococcus faecalis is a whole-genome sequenced bacterium that was isolated from a clinical specimen in 2011. This strain can be used in antimicrobial resistance research and drug development. It is resistant to vancomycin and the presence of the vanB gene was confirmed by PCR.
Catalog No. 50-238-3140