Learn More
ATCC Enterococcus faecalis (Andrewes and Horder) Schleifer and Kilpper-Balz
Supplier:  ATCC 29212/DEL
Encompass
Used in media testing, susceptibility testing, and as a CLSI quality control strain. Used as a quality control strain for BD, bioMérieux VITEK, RapID, MicroScan, and Sensititre products.
Catalog No. 50-189-136FP