Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC 51299
Encompass
Resistant to vancomycin. Sensitive to teicoplanin. CLSI quality control strain for HLAR Gentamicin and HLAR Streptomycin.
Catalog No. 50-238-4338