Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC 33186
Encompass
Enterococcus faecalis strain CN 478 is a quality control strain that can be used in susceptibility testing and the evaluation of Mueller-Hinton Agar.
Catalog No. 50-238-4294