Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC 19433
Encompass
Enterococcus faecalis strain NCTC 775 is a whole-genome sequenced bacterial type strain. It has applications in media testing, quality control, as reference material, as a control strain for Group D, and in water testing.
Catalog No. 50-238-4157