Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC 29212
Encompass
Enterococcus faecalis strain [Portland] was isolated from urine. This whole-genome sequenced bacterium has applications in food testing, media testing, quality control, and water testing.
Catalog No. 50-238-4108